MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Occurrence of aflatoxin M₁ in UHT milk and infant formula samples consumed in Burdur, Turkey.
Occurrence of aflatoxin M₁ in UHT milk and infant formula samples consumed in Burdur, Turkey.
Environmental monitoring and assessment (2014-05-31)
Fatma Sahindokuyucu Kocasari
ABSTRACT
Aflatoxins are toxic fungal metabolites found in foods and feeds. Aflatoxin M₁ (AFM₁), a metabolite of the potent carcinogen aflatoxin B₁(AFB₁), occurs in milk from animals consuming feed contaminated with AFB1. The aim of this study was to investigate the occurrence and levels of AFM₁ in ultra-high temperature (UHT) milk and infant formula samples consumed in Burdur City. During 2011, 41 samples of UHT milk and 33 samples of infant formula were randomly collected from different supermarkets and drug stores. The occurrence and contamination levels of AFM₁ in the samples were investigated using the competitive enzyme-linked immunoabsorbent assay (ELISA) method. AFM₁ was detected in 30 samples (73.2%) in concentrations ranging from 6.42 to 71.33 ng/L (mean level, 17.76 ng/L). Altogether, only three (7.3%) samples of UHT milk were contaminated at levels above the Turkish legal limit (50 ng/L). However, AFM₁ was not detected in all infant formula samples. It is concluded that despite the widespread occurrence of AFM₁ in UHT milk samples, the contamination levels were not serious for public health.
Sigma-Aldrich
Aflatoxin M1, from Aspergillus flavus
Supelco
Aflatoxin M1 solution, 0.5 μg/mL in acetonitrile, analytical standard