MilliporeSigma
  • Design and synthesis of 2α-(tetrazolylethyl)-1α,25-dihydroxyvitamin D3 as a high affinity ligand for vitamin D receptor.
Design and synthesis of 2α-(tetrazolylethyl)-1α,25-dihydroxyvitamin D3 as a high affinity ligand for vitamin D receptor.
The Journal of steroid biochemistry and molecular biology (2013-09-17)
Miki Matsuo, Asami Hasegawa, Masashi Takano, Hiroshi Saito, Shinji Kakuda, Kenichiro Takagi, Eiji Ochiai, Kyohei Horie, Midori Takimoto-Kamimura, Kazuya Takenouchi, Daisuke Sawada, Atsushi Kittaka
ABSTRACT
X-ray cocrystallographic studies of the human vitamin D receptor (hVDR)-[2α-(3-hydroxypropyl)-1α,25-dihydroxyvitamin D3 (O1C3)] complex showed that the terminal hydroxy group of the 2α-functional group of O1C3 formed a hydrogen bond with Arg274 in the ligand binding domain (LBD) of hVDR to stabilize the complex; therefore, O1C3 showed 3-times greater binding affinity for VDR than the natural hormone. Here, the effects of a heteroaromatic ring on binding to hVDR instead of the terminal OH group of O1C3 and also on preliminary biological activities were studied. We synthesized 2α-[2-(tetrazol-2-yl)ethyl]-1α,25(OH)2D3 (1a) and its regioisomer 2α-[2-(tetrazol-1-yl)ethyl]-1α,25(OH)2D3 (1b), in which 1a showed much higher hVDR binding affinity and greater osteocalcin promoter transactivation activity in human osteosarcoma (HOS) cells than those of 1b. X-ray cocrystallographic analysis of the hVDR-1a complex showed new hydrogen bond formation between one of the nitrogen atoms of the tetrazole ring and Arg274. This article is part of a Special Issue entitled '16th Vitamin D Workshop'.
Product Description
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Supelco
1α,25-Dihydroxyvitamin D3 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)