MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pegylation improves the pharmacokinetics and bioavailability of small-molecule drugs hydrolyzable by esterases: a study of phospho-Ibuprofen.
Pegylation improves the pharmacokinetics and bioavailability of small-molecule drugs hydrolyzable by esterases: a study of phospho-Ibuprofen.
The Journal of pharmacology and experimental therapeutics (2014-07-23)
George Mattheolabakis, Chi C Wong, Yu Sun, Carol A Amella, Robert Richards, Panayiotis P Constantinides, Basil Rigas
ABSTRACT
Esterase hydrolysis of drugs can accelerate their elimination, thereby limiting their efficacy. Polyethylene glycol (PEG) covalently attached to drugs (pegylation) is known to improve the efficiency of many drugs. Using as a test agent the novel phospho-ibuprofen (PI), we examined whether pegylation of PI could abrogate its hydrolytic degradation by esterases; PI, known to inhibit colon cancer growth, has a carboxylic ester hydrolyzable by carboxylesterases (CES). We covalently attached mPEG-2000 to PI (PI-PEG) and studied its stability by exposing it to cells overexpressing CES and by administering it to mice. We also evaluated PI-PEG's anticancer efficacy in human colon cancer xenografts and in Apc(min/+) mice. PI-PEG was stable in the presence of cells overexpressing CES1 or CES2, whereas PI was extensively hydrolyzed (90.2 ± 0.7%, 14.3 ± 1.1%, mean ± S.E.M.). In mice, PI was nearly completely hydrolyzed. Intravenous administration of PI-PEG resulted in significant levels in blood and in colon cancer xenografts (xenograft values in parentheses): area under the curve for 0-24 hours = 2351 (2621) (nmol/g) × h; Cmax = 1965 (886) nmol/g; Tmax = 0.08 (2) hour. The blood levels of ibuprofen, its main hydrolytic product, were minimal. Compared with controls, PI-PEG inhibited the growth of the xenografts by 74.8% (P < 0.01) and reduced intestinal tumor multiplicity in Apc(min/+) mice by 73.1% (P < 0.01), prolonging their survival (100% versus 55.1% of controls; P = 0.013). Pegylation protects PI from esterase hydrolysis and improves its pharmacokinetics. In preclinical models of colon cancer, PI-PEG is a safe and efficacious agent that merits further evaluation.
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000