MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Increased corneal hysteresis after corneal collagen crosslinking: a study based on applanation resonance technology.
Increased corneal hysteresis after corneal collagen crosslinking: a study based on applanation resonance technology.
JAMA ophthalmology (2014-08-30)
Jeannette Beckman Rehnman, Anders Behndig, Per Hallberg, Christina Lindén
ABSTRACT
A reliable tool for quantification of the biomechanical status of the cornea in conjunction with corneal collagen crosslinking (CXL) treatment is needed. To quantify the biomechanical effects of CXL in vivo. A prospective, open, case-control study was conducted at the Department of Ophthalmology, Umeå University, Umeå, Sweden. Participants included 28 patients (29 eyes) aged 18 to 28 years with progressive keratoconus and corresponding age- and sex-matched healthy individuals serving as controls. All participants were monitored during a 6-month period between October 13, 2009, and November 5, 2012. Corneal hysteresis after CXL for keratoconus. A difference in corneal hysteresis between the control group and the patients with keratoconus was found at baseline, both with an applanation resonance tonometer (ART) and an ocular response analyzer (ORA), at mean (SD) values of -1.09 (1.92) mm Hg (99% CI, -2.26 to 0.07; P = .01) and -2.67 (2.55) mm Hg (99% CI, -4.05 to -1.32; P < .001), respectively. Increased corneal hysteresis was demonstrated with an ART 1 and 6 months after CXL, at 1.2 (2.4) mm Hg (99% CI,-0.1 to 2.5; P = .02) and 1.1 (2.7) mm Hg (99% CI, -0.3 to 2.6; P = .04), respectively, but not with ORA. A decrease in corneal thickness was seen 1 and 6 months after treatment (-24 [26] µm, P < .001; and -11 [21] µm, P = .01, respectively), and a corneal flattening of -0.6 (0.7) diopters was seen at 6 months (P < .001). No significant change in intraocular pressure was identified in patients with keratoconus with any method, except for an increase at 1 month with Goldmann applanation tonometry (P = .005). To our knowledge ART is the first in vivo method able to assess the increased corneal hysteresis after CXL treatment. Given the large-scale use of CXL in modern keratoconus treatment, a tool with this capacity has a great potential value. Refinement of the ART method of measuring and quantifying corneal biomechanical properties will be a subject of further studies.
Product Description
Supelco
Riboflavin (B2), analytical standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%