MilliporeSigma
  • Exhaust and evaporative emissions from motorcycles fueled with ethanol gasoline blends.
Exhaust and evaporative emissions from motorcycles fueled with ethanol gasoline blends.
The Science of the total environment (2014-10-11)
Lan Li, Yunshan Ge, Mingda Wang, Zihang Peng, Yanan Song, Liwei Zhang, Wanli Yuan
ABSTRACT
The emission characteristics of motorcycles using gasoline and E10 (90% gasoline and 10% ethanol by volume) were investigated in this article. Exhaust and evaporative emissions of three motorcycles were investigated on the chassis dynamometer over the Urban Driving Cycle (UDC) and in the Sealed Housing for Evaporative Determination (SHED) including regulated and unregulated emissions. The regulated emissions were detected by an exhaust gas analyzer directly. The unregulated emissions including carbonyls and volatile organic compounds (VOCs) were sampled through battery-operated air pumps using tubes coated with 2,4-dinitrophenylhydrazine (DNPH) and Tenax TA, respectively. The experimental results showed that the emission factors of total hydrocarbons (THC) and carbon monoxide (CO) from E10 fueling motorcycles decreased by 26%-45% and 63%-73%, while the emission factor of NOx increased by 36%-54% compared with those from gasoline fueling motorcycles. For unregulated emissions, the emission amount of VOCs from motorcycles fueled with E10 decreased by 18%-31% while total carbonyls were 2.6-4.5 times higher than those for gasoline. For evaporative emissions of THC and VOCs, for gasoline or E10, the diurnal breathing loss (DBL) was higher than hot soak loss (HSL). Using E10 as a fuel does not make much difference in the amount of evaporative THC, while resulted in a slightly growth of 14%-17% for evaporative BETX (benzene, toluene, ethylbenzene, xylene).
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Carbon monoxide, ≥99.0%
Sigma-Aldrich
Reagent Alcohol, reagent grade
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, 94.8-95.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Toluene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, denatured, suitable for HPLC
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, denatured, denatured, reagent grade
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%