MilliporeSigma
  • Vascular-promoting therapy reduced tumor growth and progression by improving chemotherapy efficacy.
Vascular-promoting therapy reduced tumor growth and progression by improving chemotherapy efficacy.
Cancer cell (2015-01-15)
Esther Bridges, Adrian L Harris
ABSTRACT
In this issue of Cancer Cell, Wong and colleagues describe a novel approach of increasing the number of functional blood vessels in tumors using a low-dose therapy regimen of Cilengtide and Verapamil. This method enhanced gemcitabine delivery, uptake, and metabolism within tumor cells to reduce tumor growth and progression.
Product Description
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
USP
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard