MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Human heat shock protein 27 exacerbates ischemia reperfusion injury in rats by reducing the number of T regulatory cells.
Human heat shock protein 27 exacerbates ischemia reperfusion injury in rats by reducing the number of T regulatory cells.
Molecular medicine reports (2014-03-15)
Sunyi Ye, Chenxi Zhang, Jie Zhou, Jun Cheng, Zhen Lv, Lin Zhou, Haiyang Xie, Jian Wu, Shusen Zheng
ABSTRACT
Ischemia reperfusion injury (IRI) occurs in almost every liver surgery and is associated with the reduction of the liver blood flow. Ischemia impairs liver function and can even cause liver failure following surgery. The present study aimed to identify a new molecular target allowing the reduction of IRI and explore the related cellular mechanism. Adenovirus (~2.5x10(12) viral particles) bearing the human heat shock protein 27 (HSP27) gene was injected into rat liver through the ileocecal vein. Five days following the injection, ischemia was induced by clamping the median and left portal veins, hepatic arteries and bile ducts. The levels of alanine transaminase (ALT), aspartate aminotransferase (AST), glutathione (GSH) and superoxide dismutase (SOD) were measured. The infiltration of inflammatory cells and the expression of pro-inflammatory factors were investigated. The number of regulatory T cells (Tregs) was measured by flow cytometry. At 2 h following reperfusion, the group injected with HSP27 had the highest level of ALT and AST, followed by the group injected with HSP27 and treated with gadolinium trichloride (GdCl3), the empty vector-injected and the vector+GdCl3 groups. The HSP27 group also had the lowest levels of the oxidative stress-protective factors SOD and GSH, and the highest levels of pro-inflammatory factors. The number of Tregs was reduced in the groups injected with HSP27. In conclusion, the human HSP27 protein can effectively accelerate liver damage at the early stages of IRI in rats. Tregs might play a critical role in HSP27‑induced liver injury.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)