MilliporeSigma
  • Development of therapeutic polymeric nanoparticles for the resolution of inflammation.
Development of therapeutic polymeric nanoparticles for the resolution of inflammation.
Advanced healthcare materials (2014-03-25)
Suresh Gadde, Orli Even-Or, Nazila Kamaly, Apoorva Hasija, Philippe G Gagnon, Krishna H Adusumilli, Andrea Erakovic, Anoop K Pal, Xue-Qing Zhang, Nagesh Kolishetti, Jinjun Shi, Edward A Fisher, Omid C Farokhzad
ABSTRACT
Liver X receptors (LXRs) attenuate inflammation by modulating the expression of key inflammatory genes, making LXRs and their ligands particularly attractive candidates for therapeutic intervention in cardiovascular, metabolic, and/or inflammatory diseases. Herein, enhanced proresolving activity of polymeric nanoparticles (NPs) containing the synthetic LXR agonist GW3965 (LXR-NPs) is demonstrated, developed from a combinatorial library of more than 70 formulations with variations in critical physicochemical parameters. In vitro studies on peritoneal macrophages confirm that LXR-NPs are significantly more effective than the free agonist at downregulating pro-inflammatory mediators (MCP-1 and TNFα), as well as inducing the expression of LXR target genes (ABCA1 and SREBP1c). Through a zymosan-induced acute peritonitis in vivo model, LXR-NPs are found to be more efficient than free GW3965 at limiting the recruitment of polymononuclear neutrophils (50% vs 17%), suppressing the gene expression and secretion of pro-inflammatory factors MCP-1 and TNFα in peritoneal macrophages, and decreasing the resolution interval up to 4 h. Furthermore, LXR-NPs suppress the secretion of MCP-1 and TNFα by monocytes and macrophages more efficiently than the commercial drug dexamethasone. Overall, these findings demonstrate that LXR-NPs are capable of promoting resolution of inflammation and highlight the prospect of LXR-based nanotherapeutics for inflammatory diseases.
Product Description
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Ethyldiisopropylamine, ≥98.0%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
GW3965 hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Dimethyl ether, ≥99%
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Tin(II) 2-ethylhexanoate, 92.5-100.0%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material