MilliporeSigma
  • The clinical characteristics of sudden cardiac arrest in asymptomatic patients with congenital heart disease.
The clinical characteristics of sudden cardiac arrest in asymptomatic patients with congenital heart disease.
Heart and vessels (2013-11-28)
Aya Miyazaki, Heima Sakaguchi, Hideo Ohuchi, Kenji Yasuda, Nobuyuki Tsujii, Michio Matsuoka, Tetsuya Yamamoto, Satoshi Yazaki, Etsuko Tsuda, Osamu Yamada
ABSTRACT
Sudden cardiac arrest (SCA) is a major cause of death in patients with congenital heart disease (CHD). Systemic ventricular dysfunction is a reported risk factor for SCA. We retrospectively analyzed the medical records of 46 patients (age >6 years) who experienced SCA. The following underlying cardiac defects were observed: biventricular repair with affected subpulmonary right ventricle (n = 18, 39 %), biventricular repair with systemic right ventricle and Eisenmenger syndrome (n = 6 each, 13 %), Fontan circulation and unrepaired CHD (n = 5 each, 11 %), and others (n = 6, 13 %). Twenty-one patients (46 %) had no history of arrhythmias, and 21 of 43 (49 %) showed systemic ventricular ejection fraction >55 %. According to the New York Heart Association classification, 18 patients (39 %) were class I and 28 (61 %) were class II/III. SCA occurred at a younger age in class I (16 ± 5 years) than in the other classes (23 ± 10 years; P = 0.004). QRS duration was similar between the groups (136 ± 38 vs. 141 ± 50 ms; P not significant). Seven patients in class I (15 % of all SCAs) had no history of arrhythmias or features of hemodynamic abnormalities. The proportion of patients with biventricular repair and affected subpulmonary right ventricle was higher than that of patients with other defects, and the majority of SCA patients had more complicated defects than a simple repaired ventricular septal defect or an atrial septal defect. No symptoms of heart failure, history of arrhythmias, or features of hemodynamic abnormalities were observed in 15 % of the patients who experienced SCA. Prolonged QRS duration might be a predictor of SCA even in asymptomatic CHD patients. Prevention of SCA in CHD patients may require more detailed evaluation than is typically considered necessary.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material