MilliporeSigma
  • Acetylcholinesterase activity of electric eel is increased or decreased by selected monoterpenoids and phenylpropanoids in a concentration-dependent manner.
Acetylcholinesterase activity of electric eel is increased or decreased by selected monoterpenoids and phenylpropanoids in a concentration-dependent manner.
Chemico-biological interactions (2015-02-01)
María Dolores López, Francisco J Campoy, María Jesús Pascual-Villalobos, Encarnación Muñoz-Delgado, Cecilio J Vidal
ABSTRACT
The profitable insecticidal action of monoterpenoids prompted us to test their efficiency against stored-grain beetle species, via inhibition of acetylcholinesterase (AChE). For this, we first studied the ability of the monoterpenoids geraniol, linalool, camphor, fenchone, carvone and γ-terpinene, besides the phenylpropanoids trans-anethole and estragole to inhibit Electrophorus AChE. The results indicated that while AChE activity increased (15-35%) with 40 μM geraniol, camphor, γ-terpinene and linalool, the activity decreased (60-40%) with 5mM carvone, γ-terpinene, and fenchone. The Km for AChE was 0.52 ± 0.02 mM in control assays, which fell to 0.28 ± 0.01 mM or 0.32 ± 0.01 mM in assays with 20 μM linalool or γ-terpinene added. In the millimolar range, the terpenoids behaved as weak inhibitors. Unexpectedly, AChE inhibition by camphor, carvone, γ-terpinene, and fenchone gave Hill numbers ranging 2.04-1.57, supporting the idea that AChE was able to lodge more than one monoterpenoid molecule. The plots of 1/v vs. 1/S at varying monoterpenoid provided straight lines, fenchone and γ-terpinene acting as competitive inhibitors and carvone and camphor as non-competitive inhibitors. Moreover, the secondary plots of the slope KM(app)/Vmax(app) vs. [I] and of 1/Vmax(app) vs. [I] gave parabolic curves, which lent support to the proposed capacity of AChE to bind more than one monoterpenoid molecule. The fitting of the curves to a second-order polynomial equation allowed us to calculate the inhibition constants for the interaction of AChE with fenchone, γ-terpinene, carvone and camphor. The previously unnoticed increase in AChE activity with monoterpenoids should be considered as a reminder when advising the use of essential oils of plants or their constituents as anti-AChE agents to attenuate pathological signs of Alzheimer's disease.
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Geraniol, 98%
Supelco
Linalool, analytical standard
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Linalool, 97%
Supelco
trans-Anethole, analytical standard
Supelco
Geraniol, analytical standard
Sigma-Aldrich
trans-Anethole, 99%
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
(−)-Linalool, analytical standard