MilliporeSigma
  • Catalytic asymmetric hydroalkenylation of vinylarenes: electronic effects of substrates and chiral N-heterocyclic carbene ligands.
Catalytic asymmetric hydroalkenylation of vinylarenes: electronic effects of substrates and chiral N-heterocyclic carbene ligands.
Angewandte Chemie (International ed. in English) (2015-02-07)
Chun-Yu Ho, Chun-Wa Chan, Lisi He
ABSTRACT
An asymmetric tail-to-tail cross-hydroalkenylation of vinylarenes with terminal olefins was achieved by catalysis with NiH complexes bearing chiral N-heterocyclic carbenes (NHCs). The reaction provides branched gem-disubstituted olefins with high enantioselectivity (up to 94 % ee) and chemoselectivity (cross/homo product ratio: up to 99:1). Electronic effects of the substituents on the vinylarenes and on the N-aryl groups of the NHC ligands, but not a π,π-stacking mechanism, assist the steric effect and influence the outcome of the cross-hydroalkenylation.
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Styrene, analytical standard
Sigma-Aldrich
Propylene, ≥99%
Sigma-Aldrich
Styrene, SAJ first grade, ≥99.0%