MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
MicroRNA signatures of platinum-resistance in ovarian cancer.
European journal of gynaecological oncology (2015-04-16)
H C Ying, H Y Xu, J Lv, T S Ying, Q Yang
ABSTRACT
The authors utilized a microRNA (miRNA) array to compare the differentially expressed miRNAs in platinum-resistant associated ovarian cancer cells. The differential expression of microRNA between COC1 (DDP-sensitive) and platinum-resistant COC1/DDP (DDP-resistant) tumor cell lines was determined using microarray. Expression levels were further validated by real-time quantitive polymerase chain reaction (qRT-PCR). The authors identified that several miRNAs are altered in collected 86 samples of human ovarian cancer cell-lines, with four significantly deregulated miRNAs and 13 upregulated miRNAs. Of which, miR-141-3p was the most differentially expressed miRNA between COC1 group (1.7833 ± 0.7213) and COC1/DDP group (14.0433 ± 4.4895) (p < 0.05). Additionally, the product curve of PCR amplification indicated that miR-141-3p had a significant higher expression level in chemotherapy resistant group (n = 20) rather than in chemotherapy sensitive group (n = 20) (9.56 ± 1.04 vs. 1.59 ± 0.91, p < 0.05). The present results suggest that miR-141-3p might be used as a therapeutic target to modulate platinum-based chemotherapy and as a biomarker to predict chemotherapy response.
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)