MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Correlation of daytime sleepiness with urine metabolites in patients with obstructive sleep apnea.
Correlation of daytime sleepiness with urine metabolites in patients with obstructive sleep apnea.
Sleep & breathing = Schlaf & Atmung (2014-01-10)
Man-Jeong Paik, Dong-Kyu Kim, Duc-Toan Nguyen, Gwang Lee, Chae Seo Rhee, In Young Yoon, Jeong-Whun Kim
ABSTRACT
The apnea-hypopnea index (AHI) is closely related with the severity of daytime sleepiness, but excessive daytime sleepiness (EDS) is not presented on all patients with obstructive sleep apnea (OSA). It is unclear why daytime sleepiness is not always present in OSA patients even if their sleep is disrupted from the perspective of polysomnographic findings. This study aimed to analyze the correlation between sleepiness and urine metabolites of neurotransmitters involved in the arousal system. On the basis of AHI in polysomnography, 49 consecutive OSA patients were included. According to their Epworth sleepiness scale (ESS), 23 non-sleepy patients (ESS <11) and 26 sleepy patients (ESS ≥11) were included. Urine samples were collected before and after polysomnography and analyzed by gas chromatography-mass spectrometry with selective ion monitoring. Six metabolites of dopamine, norepinephrine and serotonin were analyzed. The dopamine metabolites, homovanillic acid (r = 0.366, P = 0.017) and 3,4-dihydroxyphenylacetic acid (DOPAC; r = 0.584, P < 0.0001), were significantly correlated with ESS after adjusting for age, sex, body mass index, AHI, sleep efficiency and total sleep time. A two-by-two table analysis revealed that the overnight increase in the urine DOPAC was more frequent in sleepy patients while its decrease was more frequent in non-sleepy patients (odds ratio = 3.54, P = 0.032). Urine dopamine metabolites may identify sleepy patients with OSA. In particular, the overnight change of urine DOPAC may indicate OSA patients with EDS.
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)