MilliporeSigma
  • Mussel-inspired bolaamphiphile sticky self-assemblies for the preparation of magnetic nanoparticles.
Mussel-inspired bolaamphiphile sticky self-assemblies for the preparation of magnetic nanoparticles.
Colloids and surfaces. B, Biointerfaces (2015-02-07)
Chaemyeong Lee, Sang-Yup Lee
ABSTRACT
Adopting the strong metal binding moiety of a mussel protein, a novel bolaamphiphile molecule was prepared and applied to the fabrication of magnetic core-shell nanoparticles. The novel bolaamphiphile molecule with 3,4-dihydroxyphenylalanine (DOPA) end groups was synthesized and its self-assembly was used as a template to adsorb metal ions and subsequently to produce magnetic nanoparticles. The DOPA bolaamphiphile molecule self-assembled in aqueous solution to produce nanospherical structures that exposed the catechol moiety of DOPA to the outer surface. The catechol groups adsorbed cobalt and iron ions to create magnetic metal oxide clusters on the self-assembly. Spectroscopic analysis showed that the cobalt and iron ions were coordinated with quinone, an oxidized form of the catechol. Exploiting the strong metal-adsorbing and binding properties of DOPA, dense cobalt oxide and iron oxide shell layers were created on the nanospherical self-assembly to produce magnetic core-shell nanoparticles. This study demonstrated a simple method for creating magnetic metal oxide nanoparticles that exploits the molecular binding forces and self-assembly property of DOPA.
Product Description
Supelco
Triethylamine, analytical standard
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Levodopa, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%
Sigma-Aldrich
Sodium molybdate dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium molybdate dihydrate, 99.99% trace metals basis
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, SAJ first grade, ≥98.5%
Sigma-Aldrich
Azelaic acid, 98%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Azelaic acid, technical grade, 80%
Sigma-Aldrich
Azelaic acid, technical, ~85% (GC)
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Supelco
Nitrite ion standard solution, for ion-selective electrodes, 0.1 M NO2-