MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Liver sinusoidal endothelial dysfunction after LPS administration: a role for inducible-nitric oxide synthase.
Liver sinusoidal endothelial dysfunction after LPS administration: a role for inducible-nitric oxide synthase.
Journal of hepatology (2014-07-20)
Vincenzo La Mura, Marcos Pasarín, Aina Rodriguez-Vilarrupla, Juan Carlos García-Pagán, Jaime Bosch, Juan G Abraldes
ABSTRACT
Sepsis is associated with microvascular dysfunction, which contributes to organ failure. Intrahepatic endothelial dysfunction occurs after exposure to lipopolysaccharide (LPS). The upregulation of inducible nitric oxide synthase (iNOS) has been shown to contribute to systemic vascular dysfunction after LPS administration. However, little is known about the effects of iNOS induction on the liver microcirculation. This study aimed at exploring, in the isolated rat liver perfusion model, the role of iNOS induction in liver microvascular dysfunction associated with endotoxemia. All experiments were conducted in male Wistar rats, after 24 h of LPS (5 mg/kg i.p.) or saline administration in the presence or absence of the iNOS inhibitor 1400 W (3 mg/kg i.p.), administered 3 and 23 h after LPS/saline injection. Liver microvascular function was assessed by isolated liver perfusion, followed by molecular studies and liver function tests. At 24 h, LPS induced liver endothelial dysfunction, as shown by a decreased vasodilatory response to acetylcholine and decreased eNOS phosphorylation at Ser(1176). This was associated with liver injury, assessed by an increase in liver transaminases and decreased indocyanin green clearance, and increased nitrooxidative stress. iNOS inhibition prevented liver endothelial dysfunction, blunted the development of liver injury and attenuated LPS-induced nitrooxidative stress. iNOS upregulation contributes to liver microvascular dysfunction in endotoxemia. This suggests that this mechanism deserves further exploration in studies addressing liver protection in the context of severe acute bacterial infection.
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder