MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Cytokine response in peripheral blood indicates different pathophysiological mechanisms behind anastomotic leakage after low anterior resection: a pilot study.
Cytokine response in peripheral blood indicates different pathophysiological mechanisms behind anastomotic leakage after low anterior resection: a pilot study.
Techniques in coloproctology (2014-08-26)
M B Ellebæk, G Baatrup, J Gjedsted, C Fristrup, N Qvist
ABSTRACT
Anastomotic leakage (AL) after rectosigmoid resection is a serious complication associated with high morbidity and mortality. This case-control pilot study investigated the changes in blood concentration of 10 different cytokines and 2 complement factors in relation to symptomatic AL after low anterior resection for rectosigmoid cancer. Fifty patients scheduled for resection of rectosigmoid cancer had blood samples taken the day before surgery and on post-operative days 1, 3 and 5. Four patients with symptomatic AL were identified. Twenty-two age- and disease-matched patients constituted the control group. The concentration of 10 cytokines (granulocyte macrophage colony-stimulating factor, interferon-γ, interleukin-1β, interleukin-2, interleukin-4, interleukin-5, interleukin-6, interleukin-8, interleukin-10 and tumour necrosis factor-α) and 2 complement factors (mannan-binding lectin and membrane attack complex) were measured. The present study demonstrated that plasma concentration of interleukin-1β, interleukin-6, interleukin-8 and interleukin 10 within the first 5 post-operative days was increased in patients who developed early clinical AL, whereas there were no changes in patients with late-onset AL. The demonstrated differences in the cytokine response in early and late AL may support the theory of different pathological mechanisms of AL.
Product Description
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Metronidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Metronidazole, BioXtra
Sigma-Aldrich
Metronidazole, SAJ first grade, ≥99.0%
Supelco
Metronidazole, analytical standard
Supelco
Metronidazole, VETRANAL®, analytical standard