MilliporeSigma
  • Contrast-enhanced ultrasound-guided Sentinel lymph node biopsy of the ocular conjunctiva.
Contrast-enhanced ultrasound-guided Sentinel lymph node biopsy of the ocular conjunctiva.
The Laryngoscope (2014-06-14)
Mindy R Rabinowitz, Daniel A Merton, Ji-Bin Liu, Shivam Saxena, John Pluta, John R Eisenbrey, Adam L Baker, Michael P Rabinowitz, Sara Lally, David Cognetti, Barry B Goldberg, Edmund A Pribitkin, Joseph M Curry
ABSTRACT
Sentinel lymph node biopsy (SLNB) has been utilized for cutaneous melanoma and other malignancies arising from the eye and ocular adnexa. Currently, SLNB requires blue dyes and/or radiopharmaceuticals; both of which have significant shortcomings. This study sought to evaluate the feasibility of SLNB with the use of real-time, contrast-enhanced ultrasound (CEUS) as an alternative technique for tumors arising in the conjunctiva. Prospective feasibility study in a porcine model. Twelve experiments were performed on six non-tumor-bearing Yorkshire swine. An ultrasound contrast agent, Sonazoid (GE Healthcare, Oslo, Norway), (99m) technetium ((99m) Tc), and methylene blue (MB) (Covidien, Mansfield, MA) were injected in the ocular conjunctiva. Sentinel lymph nodes (SLNs) were localized with CEUS and findings were compared to that of MB and (99m) Tc. Fisher exact test was used. Contrast-enhanced SLNs were identified within an average of 6.2 minutes from time of injection of Sonazoid. A total of 17 SLNs were identified by at least one of the three techniques. Correlation between Sonazoid and (99m) Tc was 94.1% (16/17 SLNs). Correlation between (99m) Tc and MB was 88.2% (15/17). One SLN that was positive for (99m) Tc but negative for Sonazoid and was considered to be a false positive (1/17); findings were similar for MB (1/17). Differences between the three techniques were not significant (P = .886). CEUS-guided injection of conjunctiva for SLNB is technically feasible and correlates well with standard detection techniques. This technique shows promise for rapid, real-time, intraoperative imaging for SLNB, using a widely available imaging modality and avoiding the need for radiopharmaceuticals. NA
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution