MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
HPV DNA genotyping and methylation of gene p16 INK4A in cervical LSIL.
Experimental and molecular pathology (2015-01-13)
Filomena Aste Silveira, Gutemberg Almeida, Yara Furtado, Kátia S Silva, Paula Maldonado, Silvia Cavalcanti, Maria da Gloria da Costa Carvalho
ABSTRACT
DNA methylation is the most important epigenetic change involved in the control of gene expression in human cells. Methylation of the p16(INK4a) gene occurs early in the development of cervical cancer. Low-grade squamous intraepithelial lesions (LSILs) are prevalent, and their behavior is variable. To identify the HPV DNA type, detect the methylation status of the p16(INK4A) gene, and analyze their association with the cytological evolution of LSIL over a period of two years. We conducted a cohort study with 40 participants. Cervical scrapings were collected for cytological and molecular analysis. HPV DNA detection and typing were performed by means of polymerase chain reaction (PCR) and restriction fragment length polymorphism (RFLP). Methylation-specific PCR was performed to detect methylation. HPV DNA was detected in 87% of the cases, and type 16 was the most frequent type. Methylation was detected in 11% of the cases and did not exhibit a significant correlation with the HPV type. Unfavorable cytological evolution exhibited a significant association with the presence of methylation. HPV 16 was the most frequently detected type of HPV in LSIL. Methylation of the p16(INK4A) gene was infrequent and occurred independent of the presence of HPV DNA. Methylation of the p16(INK4a) gene exhibited a significant correlation with persistence/progression of LSIL.
Product Description
Supelco
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydroquinone, JIS special grade, ≥99.0%
Sigma-Aldrich
Bromophenol Blue, JIS special grade
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
o-Xylene, analytical standard
Supelco
o-Xylene, suitable for HPLC, 98%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%