MilliporeSigma
  • Cumulative effects of the ApoE genotype and gender on the synaptic proteome and oxidative stress in the mouse brain.
Cumulative effects of the ApoE genotype and gender on the synaptic proteome and oxidative stress in the mouse brain.
The international journal of neuropsychopharmacology (2014-05-09)
Lv Shi, Xin Du, Hong Zhou, Changlu Tao, Yuntao Liu, Fantao Meng, Gao Wu, Ying Xiong, Chun Xia, Yu Wang, Guoqiang Bi, Jiang-Ning Zhou
ABSTRACT
Elderly females, particularly those carrying the apolipoprotein E (ApoE)-ε4 allele, have a higher risk of developing Alzheimer's disease (AD). However, the underlying mechanism for this increased susceptibility remains unclear. In this study, we investigated the effects of the ApoE genotype and gender on the proteome of synaptosomes. We isolated synaptosomes and used label-free quantitative proteomics, to report, for the first time, that the synaptosomal proteomic profiles in the cortex of female human-ApoE4 mice exhibited significantly reduced expression of proteins related to energy metabolism, which was accompanied by increased levels of oxidative stress. In addition, we also first demonstrated that the proteomic response in synaptic termini was more susceptible than that in the soma to the adverse effects induced by genders and genotypes. This suggests that synaptic mitochondria might be 'older' than mitochondria in the soma of neurons; therefore, they might contain increased cumulative damage from oxidative stress. Furthermore, female human-ApoE4 mice had much lower oestrogen levels in the cortex and treatment with oestrogen protected ApoE3 stable transfected C6 neurons from oxidative stress. Overall, this study reveals complex ApoE- and gender-dependent effects on synaptic function and also provides a basis for future studies of candidates based on specific pathways involved in the pathogenesis of AD. The lack of oestrogen-mediated protection regulated by the ApoE genotype led to synaptic mitochondrial dysfunction and increased oxidative stress, which might make older females more susceptible to AD.
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent