MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Alditols and monosaccharides from sorghum vinegar can attenuate platelet aggregation by inhibiting cyclooxygenase-1 and thromboxane-A2 synthase.
Alditols and monosaccharides from sorghum vinegar can attenuate platelet aggregation by inhibiting cyclooxygenase-1 and thromboxane-A2 synthase.
Journal of ethnopharmacology (2014-06-01)
Jing Li, Guoyong Yu, Junfeng Fan
ABSTRACT
Vinegar has been used as both a common seasoning and a traditional Chinese medicine. Sorghum vinegar is an excellent source of physiological substances with multiple health benefits. To evaluate the antiplatelet aggregation activity of alditols and monosaccharides extracted from sorghum vinegar and analysis its mechanism. Alditol and monosaccharide extract (AME) from sorghum vinegar was first evaluated for antiplatelet activity using the turbidimetric method. Blood was collected from healthy volunteer donors. The platelet aggregation was induced by arachidonic acid (AA), collagen, adenosine diphosphate (ADP) and thrombin in vitro. AME was divided into three experimental groups with the concentration were 0.10, 0.25 and 0.50 mg/mL. In order to determine the inhibitory activity of AME on COX1, TXS and TXA2 production experiments were conducted using the COX1, TXS and TXB2 EIA kit. Computational docking was used to find the docking pose of monosaccharides and alditols with COX1. AME showed significant induction of antiplatelet activity by arachidonic acid (AA), collagen, adenosine diphosphate (ADP) and thrombin in a concentration-dependent manner (p<0.05). AME (0.50 mg/mL) reduced the AA-induced aggregation rate to 10.35%±0.46%, which was comparable to acetylsalicylic acid (aspirin, ASA) (0.50 mg/mL, 6.35%±0.58%), a medical standard. Furthermore, AME strongly inhibited cyclooxygenase-1 (COX1) and thromboxane-A2 synthase (TXS), and subsequently attenuated thromboxane-A2 (TXA2) production. These findings indicated that AME attenuates platelet aggregation through the AA metabolism pathway. Computational docking showed that alditols (L-erythritol, L-arabitol, xylitol and D-sorbitol), monosaccharides (D-glucopyranose, D-fructofuranonse, D-xylopyranose, D-galactopyranose and D-ribose), ethyl glucoside and 3,4-(methylenedioxy) mandelic acid could dock directly into the active site of COX1. Alditols and monosaccharides from sorghum vinegar inhibit multiple steps in the platelet aggregation pathway, and may be beneficial for the treatment of cardiovascular diseases.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material