MilliporeSigma
  • BDNF-mediated modulation of glycine transmission on rat spinal motoneurons.
BDNF-mediated modulation of glycine transmission on rat spinal motoneurons.
Neuroscience letters (2014-07-06)
Jian-Dong Ding, Xian-Ye Tang, Jian-Gang Shi, Lian-Shun Jia
ABSTRACT
BDNF has a widespread distribution in the central and peripheral nervous systems, suggesting that BDNF may play a role in the regulation of motor control. However, the direct actions of BDNF on the motoneurons and their underlying mechanisms are still largely unknown to date. Therefore, by using whole-cell patch clamp recordings, quantitative RT-PCR and immunocytochemistry, the present study was designed to investigate the effects of BDNF on electrical activity and glycinergic transmission on the motoneurons and the underlying receptor mechanism. The results reveal: (i) BDNF did not produce a direct excitatory or inhibitory effect on the motoneurons; (ii) BDNF dose-dependently increased the glycinergic transmission on the motoneurons; (iii) glycinergic transmission on motoneurons was a direct postsynaptic effect; (iv) BDNF-induced enhancement of the glycinergic transmission was mediated by the activation of TrkB receptors; and (v) BDNF and its receptors TrkB had an extensive expression in the motoneurons. These results suggest that BDNF is directly involved in the regulation of glycinergic transmission on the motoneurons through postsynaptic TrkB receptors. Considering that the glycinergic synaptic transmission of motoneurons mainly comes from Renshaw cells, the important inhibitory interneurons of spinal cord, we speculate that BDNF may play an important role in the information integration in the spinal cord and participate in the sensitivity of motoneurons.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
SAFC
Glycine
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)