MilliporeSigma
IL-21 induces IL-22 production in CD4+ T cells.
Nature communications (2014-05-07)
Ada Yeste, Ivan D Mascanfroni, Meghan Nadeau, Evan J Burns, Ann-Marcia Tukpah, Andrezza Santiago, Chuan Wu, Bonny Patel, Deepak Kumar, Francisco J Quintana
ABSTRACT
Interleukin (IL)-22 produced by innate lymphoid cells (ILCs) and CD4+ T cells plays an important role in host defence and mucosal homeostasis, thus it is important to investigate the mechanisms that regulate IL-22 production. We investigated the regulation IL-22 production by CD4+ T cells. Here we show that IL-21 triggers IL-22, but not IL-17 production by CD4+ T cells. STAT3, activated by IL-21, controls the epigenetic status of the il22 promoter and its interaction with the aryl hydrocarbon receptor (AhR). Moreover, IL-21 and AhR signalling in T cells control IL-22 production and the development of dextran sodium sulphate-induced colitis in ILC-deficient mice. Thus, we have identified IL-21 as an inducer of IL-22 production in CD4+ T cells in vitro and in vivo.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard