MilliporeSigma
  • Occurrence, sources, and potential human health risks of polycyclic aromatic hydrocarbons in agricultural soils of the coal production area surrounding Xinzhou, China.
Occurrence, sources, and potential human health risks of polycyclic aromatic hydrocarbons in agricultural soils of the coal production area surrounding Xinzhou, China.
Ecotoxicology and environmental safety (2014-07-23)
Long Zhao, Hong Hou, Yuxian Shangguan, Bin Cheng, Yafei Xu, Ruifen Zhao, Yigong Zhang, Xiaozan Hua, Xiaolan Huo, Xiufeng Zhao
ABSTRACT
A comprehensive investigation of the levels, distribution patterns, and sources of polycyclic aromatic hydrocarbons (PAHs) in agricultural soils of the coal production area surrounding Xinzhou, China, was conducted, and the potential human health risks associated with the levels observed were addressed. A total of 247 samples collected from agricultural soils from the area were analyzed for sixteen PAHs, including highly carcinogenic isomers. The PAH concentrations had a range of n.d. to 782ngg(-1), with a mean value of 202ngg(-1). The two-three ring PAHs were the dominant species, making up 60 percent of total PAHs. Compared with the pollution levels and carcinogenic potential risks reported in other studies, the soil PAH concentrations in the study area were in the low to intermediate range. A positive matrix factorization model indicates that coal/biomass combustion, coal and oil combustion, and coke ovens are the primary PAH sources, accounting for 33 percent, 26 percent, and 24 percent of total PAHs, respectively. The benzo[a]pyrene equivalent (BaPeq) concentrations had a range of n.d. to 476ngg(-1) for PAH7c, with a mean value of 34ngg(-1). The BaPeq concentrations of PAH7c accounted for more than 99 percent of the ∑PAH16, which suggests that seven PAHs were major carcinogenic contributors of ∑PAH16. According to the Canadian Soil Quality Guidelines, only six of the soil samples had concentrations above the safe BaPeq value of 600ngg(-1); the elevated concentrations observed at these sites can be attributed to coal combustion and industrial activities. Exposure to these soils through direct contact probably poses a significant risk to human health as a result of the carcinogenic effects of PAHs.
Product Description
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benz[a]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benz[a]anthracene, analytical standard
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Benz[a]anthracene, 99%
Supelco
Chrysene, analytical standard
Benz[a]anthracene, BCR®, certified reference material
Chrysene, BCR®, certified reference material
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Fluoranthene, analytical standard
Sigma-Aldrich
Fluoranthene, 98%
Supelco
Benzo[b]fluoranthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Indeno[1,2,3-cd]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Indeno[1,2,3-cd]pyrene, analytical standard
Supelco
Benzo[b]fluoranthene, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Benzo[b]fluoranthene, 98%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Anthracene, analytical standard
Supelco
Pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%