MilliporeSigma
  • Facile functionalized of SBA-15 via a biomimetic coating and its application in efficient removal of uranium ions from aqueous solution.
Facile functionalized of SBA-15 via a biomimetic coating and its application in efficient removal of uranium ions from aqueous solution.
Journal of hazardous materials (2015-01-16)
Jun-Kai Gao, Li-An Hou, Guang-Hui Zhang, Ping Gu
ABSTRACT
A novel dopamine-functionalized mesoporous silica (DMS), synthesized by grafting dopamine onto a mesoporous molecular sieve (SBA-15), was developed as a sorbent to extract U(VI) from aqueous solution. The method used to modify SBA-15 was simple, facile and cost-effective. The DMS was characterized by SEM, TEM, XRD and BET, showing that the material had an ordered mesoporous structure and a large surface area. The effect of contact time, pH, ionic strength, temperature, and solid-liquid ratio on the sorption process was investigated. It was demonstrated that the adsorption of U(VI) by DMS was fast and that it can be described by the pseudo-second order-equation where the equilibrium time was 20 min. Additionally, the adsorption isotherm data were fitted well by the Langmuir model with the maximum adsorption capacity of DMS of 196 mg/g at pH 6.0. Furthermore, the influence of the K(+) and Na(+) concentrations and solid-to-liquid ratio on the sorption was very weak, and the values of the thermodynamic parameters revealed that the sorption process was exothermic and spontaneous. All the results suggested that the DMS could be used as an excellent adsorbent to remove U(VI) from aqueous solution.
Product Description
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol 5 M solution
Supelco
Dimethyl sulfide, analytical standard
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%