MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Self in vivo production of a synthetic biological drug CTLA4Ig using a minicircle vector.
Self in vivo production of a synthetic biological drug CTLA4Ig using a minicircle vector.
Scientific reports (2014-11-07)
Yeri Alice Rim, Hyoju Yi, Youngkyun Kim, Narae Park, Hyerin Jung, Juryun Kim, Seung Min Jung, Sung-Hwan Park, Ji Hyeon Ju
ABSTRACT
Cytotoxic T lymphocyte-associated antigen 4 immunoglobulin fusion protein (CTLA4Ig, abatacept) is a B7/CD28 costimulation inhibitor that can ward off the immune response by preventing the activation of naïve T cells. This therapeutic agent is administered to patients with autoimmune diseases such as rheumatoid arthritis. Its antiarthritic efficacy is satisfactory, but the limitations are the necessity for frequent injection and high cost. Minicircles can robustly express the target molecule and excrete it outside the cell as an indirect method to produce the protein of interest in vivo. We inserted the sequence of abatacept into the minicircle vector, and by successful in vivo injection the host was able to produce the synthetic protein drug. Intravenous infusion of the minicircle induced spontaneous production of CTLA4Ig in mice with collagen-induced arthritis. Self-produced CTLA4Ig significantly decreased the symptoms of arthritis. Injection of minicircle CTLA4Ig regulated Foxp3(+) T cells and Th17 cells. Parental and mock vectors did not ameliorate arthritis or modify the T cell population. We have developed a new concept of spontaneous protein drug delivery using a minicircle vector. Self in vivo production of a synthetic protein drug may be useful when biological drugs cannot be injected because of manufacturing or practical problems.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)