MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • ALK-dependent control of hypoxia-inducible factors mediates tumor growth and metastasis.
ALK-dependent control of hypoxia-inducible factors mediates tumor growth and metastasis.
Cancer research (2014-09-07)
Cinzia Martinengo, Teresa Poggio, Matteo Menotti, Maria Stella Scalzo, Cristina Mastini, Chiara Ambrogio, Elisa Pellegrino, Ludovica Riera, Roberto Piva, Domenico Ribatti, Fabio Pastorino, Patrizia Perri, Mirco Ponzoni, Qi Wang, Claudia Voena, Roberto Chiarle
ABSTRACT
Rearrangements involving the anaplastic lymphoma kinase (ALK) gene are defining events in several tumors, including anaplastic large-cell lymphoma (ALCL) and non-small cell lung carcinoma (NSCLC). In such cancers, the oncogenic activity of ALK stimulates signaling pathways that induce cell transformation and promote tumor growth. In search for common pathways activated by oncogenic ALK across different tumors types, we found that hypoxia pathways were significantly enriched in ALK-rearranged ALCL and NSCLC, as compared with other types of T-cell lymphoma or EGFR- and K-RAS-mutated NSCLC, respectively. Consistently, in both ALCL and NSCLC, we found that under hypoxic conditions, ALK directly regulated the abundance of hypoxia-inducible factors (HIF), which are key players of the hypoxia response in normal tissues and cancers. In ALCL, the upregulation of HIF1α and HIF2α in hypoxic conditions required ALK activity and its downstream signaling proteins STAT3 and C/EBPβ. In vivo, ALK regulated VEGFA production and tumor angiogenesis in ALCL and NSCLC, and the treatment with the anti-VEGFA antibody bevacizumab strongly impaired ALCL growth in mouse xenografts. Finally, HIF2α, but not HIF1α, was required for ALCL growth in vivo whereas the growth and metastasis potential of ALK-rearranged NSCLC required both HIF1α and HIF2α. In conclusion, we uncovered an ALK-specific regulation of the hypoxia response across different ALK(+) tumor types and propose HIFs as a powerful specific therapeutic target in ALK-rearranged ALCL and NSCLC.
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)