MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Selective oral ROCK2 inhibitor down-regulates IL-21 and IL-17 secretion in human T cells via STAT3-dependent mechanism.
Selective oral ROCK2 inhibitor down-regulates IL-21 and IL-17 secretion in human T cells via STAT3-dependent mechanism.
Proceedings of the National Academy of Sciences of the United States of America (2014-11-12)
Alexandra Zanin-Zhorov, Jonathan M Weiss, Melanie S Nyuydzefe, Wei Chen, Jose U Scher, Rigen Mo, David Depoil, Nishta Rao, Ben Liu, Jianlu Wei, Sarah Lucas, Matthew Koslow, Maria Roche, Olivier Schueller, Sara Weiss, Masha V Poyurovsky, James Tonra, Keli L Hippen, Michael L Dustin, Bruce R Blazar, Chuan-ju Liu, Samuel D Waksal
ABSTRACT
Rho-associated kinase 2 (ROCK2) regulates the secretion of proinflammatory cytokines and the development of autoimmunity in mice. Data from a phase 1 clinical trial demonstrate that oral administration of KD025, a selective ROCK2 inhibitor, to healthy human subjects down-regulates the ability of T cells to secrete IL-21 and IL-17 by 90% and 60%, respectively, but not IFN-γ in response to T-cell receptor stimulation in vitro. Pharmacological inhibition with KD025 or siRNA-mediated inhibition of ROCK2, but not ROCK1, significantly diminished STAT3 phosphorylation and binding to IL-17 and IL-21 promoters and reduced IFN regulatory factor 4 and nuclear hormone RAR-related orphan receptor γt protein levels in T cells derived from healthy subjects or rheumatoid arthritis patients. Simultaneously, treatment with KD025 also promotes the suppressive function of regulatory T cells through up-regulation of STAT5 phosphorylation and positive regulation of forkhead box p3 expression. The administration of KD025 in vivo down-regulates the progression of collagen-induced arthritis in mice via targeting of the Th17-mediated pathway. Thus, ROCK2 signaling appears to be instrumental in regulating the balance between proinflammatory and regulatory T-cell subsets. Targeting of ROCK2 in man may therefore restore disrupted immune homeostasis and have a role in the treatment of autoimmunity.
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.