MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Angiotensin II induces region-specific medial disruption during evolution of ascending aortic aneurysms.
Angiotensin II induces region-specific medial disruption during evolution of ascending aortic aneurysms.
The American journal of pathology (2014-07-20)
Debra L Rateri, Frank M Davis, Anju Balakrishnan, Deborah A Howatt, Jessica J Moorleghen, William N O'Connor, Richard Charnigo, Lisa A Cassis, Alan Daugherty
ABSTRACT
Angiotensin II (Ang II) promotes development of ascending aortic aneurysms (AAs), but progression of this pathology is undefined. We evaluated factors potentially involved in progression, and determined the temporal sequence of tissue changes during development of Ang II-induced ascending AAs. Ang II infusion into C57BL/6J mice promoted rapid expansion of the ascending aorta, with significant increases within 5 days, as determined by both in vivo ultrasonography and ex vivo sequential acquisition of tissues. Rates of expansion were not significantly different in LDL receptor-null mice fed a saturated fat-enriched diet, demonstrating a lack of effect of hypercholesterolemia. Augmenting systolic blood pressure with norepinephrine infusion had no significant effect on ascending aortic expansion. Pathological changes observed within 5 days of Ang II infusion included increased medial thickness and intramural hemorrhage characterized by erythrocyte extravasation in outer lamellar layers of the media. Intramedial hemorrhage was not observed after prolonged Ang II infusion, although partial medial disruption was present. Elastin fragmentation and transmural medial breaks of the ascending aorta were observed with continued Ang II infusion, which were restricted to anterior aspects. CD45(+) cells accumulated in adventitia but were minimal in media. Similar pathology was observed in tissues obtained from patients with ascending AAs. In conclusion, Ang II promotes ascending AAs through region-specific changes that are independent of hypercholesterolemia or systolic blood pressure.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications