MilliporeSigma
  • Steroids in marine aquaculture farms surrounding Hailing Island, South China: occurrence, bioconcentration, and human dietary exposure.
Steroids in marine aquaculture farms surrounding Hailing Island, South China: occurrence, bioconcentration, and human dietary exposure.
The Science of the total environment (2014-10-01)
Shan Liu, Hui Chen, Xiang-Rong Xu, Shuang-Shuang Liu, Kai-Feng Sun, Jian-Liang Zhao, Guang-Guo Ying
ABSTRACT
The occurrence, bioconcentration, and human dietary exposure via seafood consumption of 24 steroids were investigated by rapid resolution liquid chromatography-tandem mass spectrometry (RRLC-MS/MS) in six typical marine aquaculture farms surrounding Hailing Island, South China. Ten, 9, 10, 15 of 24 steroids were detected at concentrations ranging from <0.1 (testosterone) to 40 ng/L (prednisolone), from 0.1 (4-androstene-3,17-dione) to 2.4 ng/g (progesterone), from 0.3 ng/g (testosterone) to 21.4 ng/g (epi-androsterone), and from <0.1 (testosterone) to 560 ng/g (cortisol) (wet weight) in the water, sediment, feed and biota samples, respectively. Synthetic steroids (androsta-1,4-diene-3,17-dione, 17α-boldenone, 17β-boldenone, 17β-trenbolone, prednisolone, norgestrel) were detected in the feed samples, clearly demonstrating the illegal use of steroids in the feed. The field bioconcentration factors (BCFs) of steroids calculated in different aquatic organisms ranged from 93.8 to 4000. The estimated daily intakes (EDIs) of androgens, glucocorticoids, and progestagens via consumption of seafood (i.e., shrimps, crabs, mollusks, and fish) for different age groups were in the range of 33.4-134, 2061-8566, and 40.4-155 ng/d for children (2-5 years), youth (6-18 years), and adults (>18 years), respectively. Even though no significant risk from dietary exposure arises from individual steroid, elevated risk to humans can result from the occurrence of multiple steroids in the seafood raised in the aquaculture farms, especially for the sensitive populations, such as pregnant women and children.
Product Description
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%