MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Riboflavin responsive mitochondrial myopathy is a new phenotype of dihydrolipoamide dehydrogenase deficiency. The chaperon-like effect of vitamin B2.
Riboflavin responsive mitochondrial myopathy is a new phenotype of dihydrolipoamide dehydrogenase deficiency. The chaperon-like effect of vitamin B2.
Mitochondrion (2014-09-25)
Rosalba Carrozzo, Alessandra Torraco, Giuseppe Fiermonte, Diego Martinelli, Michela Di Nottia, Teresa Rizza, Angelo Vozza, Daniela Verrigni, Daria Diodato, Giovanni Parisi, Arianna Maiorana, Cristiano Rizzo, Ciro Leonardo Pierri, Stefania Zucano, Fiorella Piemonte, Enrico Bertini, Carlo Dionisi-Vici
ABSTRACT
Dihydrolipoamide dehydrogenase (DLD, E3) is a flavoprotein common to pyruvate, α-ketoglutarate and branched-chain α-keto acid dehydrogenases. We found two novel DLD mutations (p.I40Lfs*4; p.G461E) in a 19 year-old patient with lactic acidosis and a complex amino- and organic aciduria consistent with DLD deficiency, manifesting progressive exertional fatigue. Muscle biopsy showed mitochondrial proliferation and lack of DLD cross-reacting material. Riboflavin supplementation determined the complete resolution of exercise intolerance with the partial restoration of the DLD protein and disappearance of mitochondrial proliferation in the muscle. Morphological and functional studies support the riboflavin chaperon-like role in stabilizing DLD protein with rescue of its expression in the muscle.
Product Description
Supelco
Riboflavin (B2), analytical standard
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
Riboflavin, European Pharmacopoeia (EP) Reference Standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%