MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibition of human drug-metabolising cytochrome P450 and UDP-glucuronosyltransferase enzyme activities in vitro by uremic toxins.
Inhibition of human drug-metabolising cytochrome P450 and UDP-glucuronosyltransferase enzyme activities in vitro by uremic toxins.
European journal of clinical pharmacology (2014-06-24)
Kyra J Barnes, Andrew Rowland, Thomas M Polasek, John O Miners
ABSTRACT
To investigate the potential inhibitory effects of uremic toxins on the major human hepatic drug-metabolising cytochrome P450 (CYP) and UDP-glucuronosyltransferase (UGT) enzymes in vitro. Benzyl alcohol, p-cresol, indoxyl sulfate, hippuric acid and a combination of the four uremic toxins were co-incubated with human liver microsomes and selective probe substrates for the major human drug-metabolising CYP and UGT enzymes. The percentage of enzyme inhibition was calculated by measuring the rates of probe metabolite formation in the absence and presence of the uremic toxins. Kinetics studies were conducted to evaluate the K i values and mechanism(s) of the inhibition of CYP2E1, CYP3A4, UGT1A1 and UGT1A9 by p-cresol. The individual uremic toxins inhibited CYP and UGT enzymes to a variable extent. p-Cresol was the most potent individual inhibitor, producing >50% inhibition of CYP2E1, CYP3A4, UGT1A1, UGT1A9 and UGT2B7 at a concentration of 100 μM. The greatest inhibition was observed with UGT1A9. p-Cresol was shown to be an uncompetitive inhibitor of UGT1A9, with unbound K i values of 9.1 and 2.5 μM in the absence and presence of bovine serum albumin (BSA), respectively. K i values for p-cresol inhibition of human liver microsomal CYP2E1, CYP3A4 and UGT1A1 ranged from 43 to 89 μM. A combination of the four uremic toxins produced >50% decreases in the activities of CYP1A2, CYP2C9, CYP2E1, CYP3A4, UGT1A1, UGT1A9 and UGT2B7. Uremic toxins may contribute to decreases in drug hepatic clearance in individuals with kidney disease by inhibition of hepatic drug-metabolising enzymes.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
USP
Phenytoin, United States Pharmacopeia (USP) Reference Standard
USP
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Phenytoin, Pharmaceutical Secondary Standard; Certified Reference Material
Phenytoin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration), meets EP testing specifications
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Sigma-Aldrich
Benzyl alcohol, SAJ special grade, ≥99.0%
Supelco
Codeine monohydrate, analytical standard
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
Sigma-Aldrich
Sulfaphenazole, ≥98%
Supelco
Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Codeine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Supelco
Benzyl alcohol, analytical standard