MilliporeSigma
  • Synthesis of g-C3N4/Fe3O4 nanocomposites and application as a new sorbent for solid phase extraction of polycyclic aromatic hydrocarbons in water samples.
Synthesis of g-C3N4/Fe3O4 nanocomposites and application as a new sorbent for solid phase extraction of polycyclic aromatic hydrocarbons in water samples.
Talanta (2014-12-06)
Man Wang, Shihai Cui, Xiaodi Yang, Wentao Bi
ABSTRACT
An easy preparation of g-C3N4/Fe3O4 nanocomposites by chemical co-precipitation has been demonstrated. The as-prepared materials were characterized by X-ray diffraction, transmission electron microscopy, Fourier transform infrared spectroscopy and thermogravimetric analysis. The high affinity of g-C3N4 toward polycyclic aromatic hydrocarbons and the magnetic behavior of Fe3O4 were combined to provide an efficient and simple magnetic solid phase extraction (MSPE). The adsorption and desorption of polycyclic aromatic hydrocarbons on g-C3N4/Fe3O4 were examined. Different factors affecting the magnetic solid phase extraction of polycyclic aromatic hydrocarbons were assessed in terms of adsorption, desorption, and recovery. Under the optimized conditions, the proposed method showed good limits of detection (LOD, S/N=3) in the range of 0.05-0.1 ng mL(-1) and precision in the range of 1.8-5.3% (RSDs, n=3). This method was also successfully applied to the analysis of real water samples; good spiked recoveries over the range of 80.0-99.8% were obtained.
Sigma-Aldrich
Phenanthrene, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Benz[a]anthracene, 99%
Benz[a]anthracene, BCR®, certified reference material
Sigma-Aldrich
Fluoranthene, 98%
Supelco
Melting point standard 79-81°C, analytical standard
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)
Supelco
Benz[a]anthracene, analytical standard
Supelco
Phenanthrene, analytical standard, for environmental analysis
Supelco
Fluoranthene, analytical standard
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Supelco
Benz[a]anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Iron(II) chloride tetrahydrate, SAJ first grade, 98.0-102.0%
Supelco
Phenanthrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%
Pyrene, BCR®, certified reference material
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Acenaphthylene, 99%
Sigma-Aldrich
Melamine, 99%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Acenaphthylene, 75%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Sigma-Aldrich
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)