MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Selective retention of basic compounds by metal aquo-ion affinity chromatography.
Selective retention of basic compounds by metal aquo-ion affinity chromatography.
Journal of separation science (2014-07-22)
Yoshiki Asakawa, Eiichi Yamamoto, Naoki Asakawa
ABSTRACT
A novel metal aquo-ion affinity chromatography has been developed for the analysis of basic compounds using heat-treated silica gel containing hydrated metal cations (metal aquo-ions) as the packing material. The packing materials of the metal aquo-ion affinity chromatography were prepared by the immobilization of a single metal component such as Fe(III), Al(III), Ag(I), and Ni(II) on silica gel followed by extensive heat treatment. The immobilized metals form aquo-ions to present cation-exchange ability for basic analytes and the cation-exchange ability for basic analytes depends on pKa of the immobilized metal species. In the present study, to evaluate the retention characteristics of metal aquo-ion affinity chromatography, the on-line solid-phase extraction of drugs was investigated. Obtained data clearly evidence the selective retention capability of metal aquo-ion affinity chromatography for basic analytes with sufficient capacity.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Millipore
TDA Reagent, suitable for microbiology
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Aluminum chloride hexahydrate, meets EP, BP, USP testing specifications
Sigma-Aldrich
Aluminum chloride hexahydrate, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%