MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Antiulcerogenic activity of Carica papaya seed in rats.
Naunyn-Schmiedeberg's archives of pharmacology (2014-11-25)
Lorraine Aparecida Pinto, Kátia Wolff Cordeiro, Viviane Carrasco, Carlos Alexandre Carollo, Cláudia Andréa Lima Cardoso, Eliana Janet Sanjinez Argadoña, Karine de Cássia Freitas
ABSTRACT
The purpose of the present study was to evaluate the gastroprotective and healing effects of the methanolic extract of the seed of the papaya Carica papaya L. (MECP) in rats. Models of acute gastric ulcer induction by ethanol and indomethacin and of chronic ulcer by acetic acid were used. The gastric juice and mucus parameters were evaluated using the pylorus ligation model, and the involvement of sulfhydryl compounds (GSH) and nitric oxide in the gastroprotective effect was analyzed using the ethanol model. The toxicity was assessed through toxicity tests. No signs of toxicity were observed when the rats received a single dose of 2000 mg/kg of extract. The MECP in doses of 125, 250, and 500 mg/kg significantly reduced the gastric lesion with 56, 76, and 82 % inhibition, respectively, and a dose of 30 mg/kg lansoprazole showed 79 % inhibition in the ethanol model. MECP (125, 250, 500 mg/kg) and cimetidine (200 mg/kg) reduced the gastric lesion in the indomethacin model, with 62, 67, 81, and 85 % inhibition, respectively. The MECP (500 mg/kg) and cimetidine (200 mg/kg) treatments showed a reduction in ulcerative symptoms induced by acetic acid by 84 and 73 %, respectively. The antiulcerogenic activity seems to involve GSH because the inhibition dropped from 72 to 13 % in the presence of a GSH inhibitor. Moreover, the MECP showed systemic action, increasing the mucus production and decreasing gastric acidity. Treatments with MECP induce gastroprotection without signs of toxicity. This effect seems to involve sulfhydryl compounds, increased mucus, and reduced gastric acidity.
Product Description
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%