MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Exposure evaluation of adult male Japanese smokers switched to a heated cigarette in a controlled clinical setting.
Exposure evaluation of adult male Japanese smokers switched to a heated cigarette in a controlled clinical setting.
Regulatory toxicology and pharmacology : RTP (2014-05-14)
Chikako Sakaguchi, Aoi Kakehi, Naoki Minami, Akira Kikuchi, Yasuyuki Futamura
ABSTRACT
The objective of this clinical study was to investigate changes in levels of biomarkers of exposure (BOEs) in healthy Japanese male smokers who switched to a prototype heated cigarette (HC). This was a controlled, semi-randomized, open-label, residential study conducted in Japan. A total of 70 healthy Japanese male smokers were enrolled. Following enrollment, subjects smoked their usual brand of cigarette for 2days and were subsequently randomized either to an HC group or a 10mg tar conventional cigarette (CC10) group for four consecutive weeks. Levels of BOEs for ten selected cigarette smoke constituents (nicotine, carbon monoxide (CO), benzene, 1,3-butadiene, acrolein, hydrogen cyanide, crotonaldehyde, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone [NNK], pyrene, 4-aminobiphenyl), and urine mutagenicity were measured at several time points during the study period. At the end of the study period, except for blood carboxyhemoglobin, levels of BOEs for the other nine constituents and urine mutagenicity were significantly lower in the HC group compared to the CC10 group. These results suggest that exposure to most cigarette smoke constituents, except CO, can be reduced by switching from CC10 to HC.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
2-Propanol, JIS special grade, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Propanol, suitable for HPLC
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥97%
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG