MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Development of a liquid chromatography-tandem mass spectrometry method for the determination of Grayanotoxins in rat blood and its application to toxicokinetic study.
Development of a liquid chromatography-tandem mass spectrometry method for the determination of Grayanotoxins in rat blood and its application to toxicokinetic study.
Biomedical chromatography : BMC (2014-05-02)
Hwang Eui Cho, Su Youn Ahn, Dong-Woo Kim, Sang-Hee Woo, Seung-Hyeok Park, Kyunghwa Hwang, Dong-Cheul Moon, Suncheun Kim
ABSTRACT
A sensitive and specific high-performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was developed for the determination of Grayanotoxin I (GTX I) and Grayanotoxin III (GTX III) in rat whole blood. Grayanotoxins (GTXs) and clindamycin as internal standard (IS) were extracted from rat blood via solid-phase extraction using PEP solid-phase extraction cartridges. Chromatographic separation of the analytes was achieved on a Kinetex C18 (100 × 2.1 mm, 2.6 µm) reversed-phase column using a gradient elution with the mobile phase of 1% acetic acid in water and methanol at a flow rate of 0.2 mL/min. Electrospray ionization mass spectrometry was operated in the positive ion mode with multiple reaction monitoring. The calibration curves obtained were linear over the concentration range of 1-100 ng/mL with a lower limit of quantification of 1 ng/mL for GTXs. The relative standard deviation of intra-day and inter-day precision was below 6.8% and accuracy ranged from 94.8 to 106.6%. The analytes were stable in the stability studies. The validated method was successfully applied to the quantification and toxicokinetic study of GTXs in rats for the first time after oral administration of 11.52 mg/kg mad honey and 0.35 mg/kg GTX III, respectively.
Product Description
Supelco
Ethyl acetate, analytical standard
Supelco
Clindamycin hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Clindamycin hydrochloride, lincosamide antibiotic
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Clindamycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Clindamycin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis