MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The three most common human papillomavirus oncogenic types and their integration state in Thai women with cervical precancerous lesions and carcinomas.
The three most common human papillomavirus oncogenic types and their integration state in Thai women with cervical precancerous lesions and carcinomas.
Journal of medical virology (2014-08-06)
Sirinart Aromseree, Arkom Chaiwongkot, Tipaya Ekalaksananan, Bunkerd Kongyingyoes, Natcha Patarapadungkit, Chamsai Pientong
ABSTRACT
To understand the potential role in cervical cancer development of the three most common high-risk human papillomavirus (HR-HPVs) in Thai women, HPV genotypes and viral genome statuses in different cervical lesions were investigated. Cervical tissues consisting of no cervical intraepithelial neoplasia (84 cases), grade I cervical intraepithelial neoplasia (176 cases), grade II-III cervical intraepithelial neoplasia (91 cases), and squamous cell carcinoma (66 cases) were subjected for HPV genotyping by polymerase chain reaction (PCR) and reverse line blot hybridization assay and for HPV genome status determination by amplification of papillomavirus oncogene transcripts (APOT) assay. HPV prevalence was 28.6% in no cervical intraepithelial neoplasia, 40.3% in grade I cervical intraepithelial neoplasia, 70.3% in grade II-III cervical intraepithelial neoplasia and 86.4% in squamous cell carcinoma cases. The three most common HR-HPV types were HPV 16, 58, and 18 which were distributed in all cervical lesions. HPV physical statuses could be investigated in 4 no cervical intraepithelial neoplasias, 2 grade I cervical intraepithelial neoplasias, 28 grade II-III cervical intraepithelial neoplasias and 31 squamous cell carcinomas. The integrated-derived transcripts were found 3.6% in grade II-III cervical intraepithelial neoplasia and 48.4% in squamous cell carcinoma, whereas no viral genome integration was found in the group of no cervical intraepithelial neoplasia or grade I cervical intraepithelial neoplasia samples. The frequencies of HR-HPV integration in squamous cell carcinoma were found 40%, 100%, 20% of HPV 16, 18, and 58. This study indicates the oncogenic potential ability of the three most common HR-HPVs associated with cervical cancer progression.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)