MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Radioimmunoassay of human fibrinopeptide B and kinetics of fibrinopeptide cleavage by different enzymes.
Radioimmunoassay of human fibrinopeptide B and kinetics of fibrinopeptide cleavage by different enzymes.
The Journal of clinical investigation (1975-08-01)
S B Bilezikian, H L Nossel, V P Butler, R E Canfield
PMID50328
ABSTRACT
Thrombin converts fibrinogen to fibrin monomer by cleaving fibrinopeptides A and B (FPA and FPB) from the amino terminal ends of the A (alpha) and B (beta) chains. A radioimmunoassay capable of measuring the A peptide in human blood as an index of thrombin action in vivo has been described previously. This paper describes the development of a radioimmunoassay for FPB and the use of both assays in the demonstration of distinctive patterns of cleavage of the amino terminal ends of the A (alha) and B (beta) chains of fibrinogen by various enzymes. Antisera were raised in rabbits to a synthetic analogue of FPB coupled to bovine serum albumin. FPB analogue was couple to desaminotyrosine and radiolabeled with 125I by the chloramine-T technique. The radiolabeled peptide was bound by the antiserum, and binding was inhibited by synthetic or native FPB. Unbound tracer was separated from bound tracer by charcoal adsorption. The senistivity of the assay was such that 50% inhibition of binding of the tracer was caused by 1.25 ng of the native FPB. Fibrinogen was treated with thrombin, plasmin, trypsin, Reptilase, and an extract of the venom from Ancistrodon contortrix contortrix (ACC). After ethanol precipitation and centrifugation, dialysates of enzymatically altered fibrinogen were assayed for FPA and FPB. The action of thrombin on fibrinogen resulted in a rapid release of FPA and a slower release of FPB. Plasmin cleaved a segment(s) of the B (beta) chain which included FPB but cleaved no detectable FPA-containing material for the first 2 h of incubation. In the case of plasmin-treated fibrinogen, the dialysates had been further treated with thrombin before being assayed for FPA and FPB. Trypsin rapidly cleaved both peptides, the B before the A. Reptilase cleaved only FPA in 24 h. ACC cleaved FPB at a rapid rate, with a slowere cleavage of FPA. The distinctive cleavage patterns produced by the serine proteases may be useful in interpreting the levels of FPA and FPB measured in human blood and in studying the generation of FPA and FPB in clinical blood samples.
Sigma-Aldrich
[Glu1]-Fibrinopeptide B human, ≥90% (HPLC)