MilliporeSigma
  • NIPA defines an SCF-type mammalian E3 ligase that regulates mitotic entry.
NIPA defines an SCF-type mammalian E3 ligase that regulates mitotic entry.
Cell (2005-07-13)
Florian Bassermann, Christine von Klitzing, Silvia Münch, Ren-Yuan Bai, Hiroyuki Kawaguchi, Stephan W Morris, Christian Peschel, Justus Duyster
ABSTRACT
The regulated oscillation of protein expression is an essential mechanism of cell cycle control. The SCF class of E3 ubiquitin ligases is involved in this process by targeting cell cycle regulatory proteins for degradation by the proteasome, with the F-box subunit of the SCF specifically recruiting a given substrate to the SCF core. Here we identify NIPA (nuclear interaction partner of ALK) as a human F-box-containing protein that defines an SCF-type E3 ligase (SCF(NIPA)) controlling mitotic entry. Assembly of this SCF complex is regulated by cell-cycle-dependent phosphorylation of NIPA, which restricts substrate ubiquitination activity to interphase. We show nuclear cyclin B1 to be a substrate of SCF(NIPA). Inactivation of NIPA by RNAi results in nuclear accumulation of cyclin B1 in interphase, activation of cyclin B1-Cdk1 kinase activity, and premature mitotic entry. Thus, SCF(NIPA)-based ubiquitination may regulate S-phase completion and mitotic entry in the mammalian cell cycle.
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)