MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Monitoring the inorganic chemical reaction by surface-enhanced Raman spectroscopy: A case of Fe³⁺ to Fe²⁺ conversion.
Monitoring the inorganic chemical reaction by surface-enhanced Raman spectroscopy: A case of Fe³⁺ to Fe²⁺ conversion.
Talanta (2015-12-24)
Suhua Qin, Juan Meng, Xianghu Tang, Liangbao Yang
ABSTRACT
Monitoring the process of organic chemical reactions to study the kinetics by surface-enhanced Raman spectroscopy (SERS) is currently of immense interest. However, monitoring the inorganic chemical reaction is still an extremely difficulty for researchers. This study exactly focused on the monitor of inorganic chemical reaction. Capillary coated with silver nanoparticles was introduced, which was an efficient platform for monitoring reactions with SERS due to the advantages of sensitivity and excellent reproducibility. The photoreduction of [Fe(phen)3](3+) to [Fe(phen)3](2+) was used as model reaction to demonstrated the feasibility of SERS monitoring inorganic chemical reaction by involving in metal-organic complexes. Moreover, the preliminary implementation demonstrated that the kinetics of photoreduction can be real-time monitored by in situ using the SERS technique on a single constructed capillary, which may be useful for the practical application of SERS technique.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Acetone, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, suitable for HPLC