MilliporeSigma
  • Botanical features for identification of Gymnosporia arenicola dried leaf.
Botanical features for identification of Gymnosporia arenicola dried leaf.
Microscopy research and technique (2015-08-26)
Gustavo Da Silva, Rita Serrano, Elsa Teixeira Gomes, Olga Silva
ABSTRACT
Gymnosporia arenicola Jordaan (Celastraceae) is a shrub or small tree, which naturally occurs in coastal sand dunes of Southern Mozambique and South Africa. Its dried leaf is often used in traditional medicine for the treatment of infectious and inflammatory diseases. Hereby, we present results of studies carried out according to the pharmacopoeia standards for the identification of herbal drugs, in the whole, fragmented, and powdered plant material. These results were complemented with scanning electron microscopy and histochemical techniques. The leaf microscopic analysis revealed a typical dorsiventral mesophyll with a corresponding spongy parenchyma-palisade parenchyma ratio of 0.60, anomocytic and paracytic stomata, papillate cells with a diameter of 4.00 ± 0.40 µm, multicellular uniseriate nonglandular trichomes with a length of 27.00 ± 4.10 µm and cristalliferous idioblasts containing calcium oxalate cluster crystals with a diameter of 23.04 ± 5.84 µm. The present findings demonstrate that the G. arenicola leaf has both nonglandular trichomes and hypoderm, features not previously described in the corresponding botanical section (Gymnosporia sect. Buxifoliae Jordaan). The establishment of these new botanical markers for the identification of G. arenicola leaf is essential for quality, safety and efficacy reasons.
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, JIS special grade
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%