MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Alkaloids from the bulbs of Lycoris longituba and their neuroprotective and acetylcholinesterase inhibitory activities.
Alkaloids from the bulbs of Lycoris longituba and their neuroprotective and acetylcholinesterase inhibitory activities.
Archives of pharmacal research (2014-09-16)
Yun-Yun Zhu, Xue Li, Heng-Yi Yu, Yu-Fang Xiong, Peng Zhang, Hui-Fang Pi, Han-Li Ruan
ABSTRACT
Three novel alkaloids (1-3), together with nineteen known ones (4-22), were isolated from the bulbs of Lycoris longituba. Their structures were elucidated on the basis of extensive spectroscopic analyses, which belong to several Amaryllidaceae alkaloid skeletons. Among them, the harmane-type alkaloids (the new compound 1 and the known compounds 5, 6 and 7) were found for the first time from Lycoris genus. The isolates were tested for their neuroprotective activities against CoCl2, H2O2 and Aβ25-35-induced SH-SY5Y cell injuries, and the majority of them exhibited neuroprotective activities of different degrees. The acetylcholinesterase (AChE) inhibitory activities of the isolated alkaloids were also evaluated, while compounds 12, 14-20 and 22 exhibited extremely significant AChE inhibitory activities.
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)