MilliporeSigma
  • Inhibition of DMBA-induced Oral Squamous Cells Carcinoma Growth by Brazilian Red Propolis in Rodent Model.
Inhibition of DMBA-induced Oral Squamous Cells Carcinoma Growth by Brazilian Red Propolis in Rodent Model.
Basic & clinical pharmacology & toxicology (2015-01-06)
Danielle R Ribeiro, Ângela Valéria F Alves, Esaú P dos Santos, Francine F Padilha, Margarete Z Gomes, Alessandra S Rabelo, Juliana C Cardoso, Adna Prado Massarioli, Severino Matias de Alencar, Ricardo Luiz C de Albuquerque-Júnior
ABSTRACT
We investigated the effect of oral administration of hydroalcoholic extract of Brazilian red propolis (HERP) on DMBA-induced oral squamous cell carcinomas (OSCC) in rodents. The chemical components of the HERP were assessed by high-performance liquid chromatography (HPLC). Carcinogenesis was topically induced in the lower lip of 25 rats using 9,10-dimethyl-1,2-benzanthracene (DMBA); the tumour was treated with saline (TUM1) and Tween 80 (TUM2) as well as HERP at 10, 50 and 100 mg/kg (HERP10, HERP50 and HERP100, respectively) for 20 weeks. Topical application of saline and oral administration of 100 mg/kg HERP was used in five rats as a control group (CTR). After 26 weeks, the histological malignancy grading and immunohistochemical expression of Ki-67 and p16(INK4A) were assessed in the tumours/tissue samples. The compounds identified were propyl gallate, daidzein, catechin, epicatechin, formononetin and biochanin A. Formononetin, daidzein and biochanin A showed concentration of 23.29, 0.38 and 0.67 mg/g of HERP, respectively. HERP at doses of 50 and 100 mg/kg inhibited 40% of OSCC growth and promoted a 3-week delay in development of clinically detectable tumours. Epithelial dysplasia was observed in all samples with no clinical tumour, except in CTR. No significant difference in the immunoexpression of Ki-67 and p16(INK4A) was observed between HERP-treated and saline/Tween 80-treated groups (p > 0.05). Our results suggest that HERP exerts chemopreventive activity on the progression of DMBA-induced epithelial dysplasia to OSCC in an experimental model of labial carcinogenesis; however, this effect is not associated with Ki-67 and p16(INK4A) immunoexpression.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%