- High Efficiency Hybrid Solar Cells Using Nanocrystalline Si Quantum Dots and Si Nanowires.
High Efficiency Hybrid Solar Cells Using Nanocrystalline Si Quantum Dots and Si Nanowires.
ACS nano (2015-07-15)
Mrinal Dutta, Lavanya Thirugnanam, Pham Van Trinh, Naoki Fukata
PMID26167772
ABSTRACT
We report on an efficient hybrid Si nanocrystal quantum dot modified radial p-n junction thinner Si solar cell that utilizes the advantages of effective exciton collection by energy transfer from nanocrystal-Si (nc-Si) quantum dots to underlying radial p-n junction Si nanowire arrays with excellent carrier separation and propagation via the built-in electric fields of radial p-n junctions. Minimization of recombination, optical, and spectrum losses in this hybrid structure led to a high cell efficiency of 12.9%.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis