MilliporeSigma
Interactions of Isophorone Derivatives with DNA: Spectroscopic Studies.
PloS one (2015-06-13)
Marco Deiana, Katarzyna Matczyszyn, Julien Massin, Joanna Olesiak-Banska, Chantal Andraud, Marek Samoc
ABSTRACT
Interactions of three new isophorone derivatives, Isoa Isob and Isoc with salmon testes DNA have been investigated using UV-Vis, fluorescence and circular dichroism spectroscopic methods. All the studied compounds interact with DNA through intercalative binding mode. The stoichiometry of the isophorone/DNA adducts was found to be 1:1. The fluorescence quenching data revealed a binding interaction with the base pairs of DNA. The CD data indicate that all the investigated isophorones induce DNA modifications.
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %