MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Associations between tumor markers and the risk of colorectal polyp recurrence in Chinese people.
Associations between tumor markers and the risk of colorectal polyp recurrence in Chinese people.
International journal of clinical and experimental medicine (2015-07-02)
Jing Tong, Ying Wang, Bing Chang, Dai Zhang, Bingyuan Wang
ABSTRACT
Colorectal cancer primarily arises from the polyps of the colon. Early identification of the recurrence of colorectal polyps represents the best opportunity to prevent the occurrence of colorectal cancer. Thus, the high risk of recurrence is the primary problem. The aim of this study was to examine the associations between tumor markers (CEA and CA19-9) and colorectal polyp recurrence in Chinese people. The risk of colorectal polyp recurrence was studied in 156 subjects (113 males and 43 females, 58.654±11.447 years old) who underwent colonoscopy and polypectomy for the first time between January 2011 and August 2014 at the First Affiliated Hospital of China Medical University, Shenyang, Liaoning, China. All subjects underwent colonoscopies within 6-12 months. Between the colorectal polyp recurrence group and the no recurrence group, the CEA and CA19-9 levels were significantly different, and the trend analyses were consistent with increased risks of recurrence with increasing CEA and CA19-9 levels among the males and patients with single polyps, multiple polyps and proximal colon polyps. The recurrence of colorectal polyps was significantly associated with increased CEA and CA19-9 levels.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications