MilliporeSigma
  • Carotid plaque inflammation assessed with (18)F-FDG PET/CT is higher in symptomatic compared with asymptomatic patients.
Carotid plaque inflammation assessed with (18)F-FDG PET/CT is higher in symptomatic compared with asymptomatic patients.
International journal of stroke : official journal of the International Stroke Society (2015-01-16)
Karolina Skagen, Kjersti Johnsrud, Kristin Evensen, Helge Scott, Kirsten Krohg-Sørensen, Frode Reier-Nilsen, Mona-Elisabeth Revheim, Jan Gunnar Fjeld, Mona Skjelland, David Russell
ABSTRACT
Carotid artery plaque inflammation is thought to be an important marker of plaque vulnerability and increased stroke risk. The main aim of this study was to assess the level of agreement between 2-deoxy-2-[(18)F] fluoro-D-glucose (18F-FDG) uptake on PET (positron emission tomography) scan in carotid plaques, with cerebrovascular symptoms, carotid plaque ultrasound echogenicity and histological assessments of plaque inflammation. Thirty-six patients with ≥70% carotid stenosis scheduled for carotid endarterectomy underwent a Colour Duplex ultrasound, (18)F-FDG PET/CT and blood tests less than 24 h prior to surgery. Plaques were defined as symptomatic when associated with ipsilateral cerebral ischemic symptoms within 30 days prior to inclusion. Plaques were assessed histologically following endarterectomy. The level of agreement between (18)F-FDG uptake (mean SUVmax and SUVmax ), and target-to-background ratio, symptoms, plaque echolucency, and histological evidence of inflammation was assessed. The amount of (18)F-FDG uptake in plaques and the amount of inflammation on histological assessment were significantly correlated (r = 0·521, P = 0·003). (18)F-FDG uptake was significantly higher in symptomatic plaques with median SUVmax 1·75 (1·26-2·04) in symptomatic, and 1·43 (1·15-2·28) in asymptomatic patients (P = 0·03). (18)F-FDG uptake was also positively correlated with echolucency on Doppler ultrasound (P = 0·03). (18)F-FDG uptake on PET/CT correlated with histological assessments of inflammation and was higher in patients with symptomatic compared with asymptomatic carotid artery plaques. These results support the use of (18)F-FDG PET/CT in the detection inflammation in carotid atherosclerosis, which may be of help in the detection of vulnerable plaques.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Fluorescein di(β-D-galactopyranoside)
Tantalum(V) ethoxide, packaged for use in deposition systems