MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Coronary severity score and C-reactive protein predict major adverse cardiovascular events in patients with stable coronary artery disease (from the Taichung CAD study).
Coronary severity score and C-reactive protein predict major adverse cardiovascular events in patients with stable coronary artery disease (from the Taichung CAD study).
Clinica chimica acta; international journal of clinical chemistry (2015-03-31)
Hung-Chih Pan, Wayne H-H Sheu, Wen-Jane Lee, Wen-Lieng Lee, Ying-Chieh Liao, Kuo-Yang Wang, I-Te Lee, Jun-Sing Wang, Kae-Woei Liang
ABSTRACT
Whether angiographic coronary severity really predicts future major adverse cardiovascular events (MACEs) in patients with coronary artery disease (CAD) is uncertain. Few studies have compared the efficacy of SYNTAX, Gensini and Jeopardy scores in predicting MACE in stable CAD. We collected data of MACE, including all-cause mortality, all strokes, new myocardial infarction and unplanned repeat revascularization, in subjects with stable CAD from our catheterization databank. Coronary severity was graded with SYNTAX, Gensini and Jeopardy scoring systems. During a median follow-up period of 42months, 39 out of the 181 subjects developed at least 1 MACE. Those with MACE had a significantly higher baseline high sensitivity C-reactive protein (hs-CRP) (p=0.025). Multivariate analysis showed that coronary severity score, hs-CRP and diabetes mellitus were significant predictors for MACE. Kaplan-Meier estimates showed a significant difference in MACE-free rates between SYNTAX binary scores (≥15 vs. <15, p=0.043), Gensini binary scores (≥36 vs. <36, p=0.048) and Jeopardy binary scores (≥4 vs. <4, p=0.001). Coronary severity score, hs-CRP and diabetes mellitus independently predicted MACE in patients with stable CAD. The Jeopardy score is simple to calculate and as effective for predicting MACE in stable CAD as the complex SYNTAX score.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications