MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Small-nerve-fiber pathology in critical illness documented by serial skin biopsies.
Small-nerve-fiber pathology in critical illness documented by serial skin biopsies.
Muscle & nerve (2014-10-14)
Miroslav Skorna, Roman Kopacik, Eva Vlckova, Blanka Adamova, Milena Kostalova, Josef Bednarik
ABSTRACT
Small-fiber pathology can develop in the acute phase of critical illness and may explain chronic sensory impairment and pain in critical care survivors. Eleven adult ischemic stroke patients in a neurocritical care unit were enrolled in an observational cohort study. Intraepidermal nerve fiber density (IENFD) in the distal leg was assessed on admission to the intensive care unit and 10-14 days later, together with electrophysiological testing. Of the 11 patients recruited, 9 (82%) had sepsis or multiple-organ failure. Median IENFD on admission (5.05 fibers/mm) decreased significantly to 2.18 fibers/mm (P < 0.001), and abnormal IENFD was found in 6 patients (54.5%). Electrodiagnostic signs of large-fiber neuropathy and/or myopathy were found in 6 patients (54.5%), and autonomic dysfunction was found in 2 patients (18.2%). Serial IENFD measurements confirmed the development of small-fiber sensory involvement in the acute phase of critical illness.
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 12 M
Sigma-Aldrich
Hydrochloric acid solution, 0.05 M
Sigma-Aldrich
Hydrochloric acid solution, 2 M
Sigma-Aldrich
Hydrochloric acid solution, 1 M
Sigma-Aldrich
Hydrochloric acid solution, 0.5 M
Sigma-Aldrich
Hydrochloric acid solution, 6 M
Sigma-Aldrich
Hydrogen chloride – ethanol solution, 0.1 M in ethanol
Sigma-Aldrich
Hydrochloric acid solution, 0.2 M
Sigma-Aldrich
Hydrochloric acid solution, 0.02 M
Sigma-Aldrich
Hydrochloric acid solution, 0.01 M