MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Disruption of exogenous eGFP gene using RNA-guided endonuclease in bovine transgenic somatic cells.
Disruption of exogenous eGFP gene using RNA-guided endonuclease in bovine transgenic somatic cells.
Zygote (Cambridge, England) (2014-11-27)
WooJae Choi, SooYoung Yum, SongJeon Lee, WonWu Lee, JiHyun Lee, SeokJoong Kim, OkJae Koo, ByeongChun Lee, Goo Jang
ABSTRACT
Genome-editing technologies are considered to be an important tool for generating gene knockout cattle models. Here, we report highly efficient disruption of a chromosomally integrated eGFP gene in bovine somatic cells using RNA-guided endonucleases, a new class of programmable nucleases developed from a bacterial Clustered Regularly Interspaced Short Palindromic Repeats (CRISPR)/Cas9 system. In the present study, we obtained homogenously eGFP-expressing primary fibroblasts from cloned bovine transgenic embryonic tissues and employed them for further analysis. CRISPR/Cas9 plasmids specifically targeting the eGFP gene were transfected into the eGFP fibroblasts by electroporation. After 10 days of culture, more than 40% of the cells had lost eGFP expression in fluorescence activated cell sorting (FACS) analysis. Targeted sequences of the transfected cells were analyzed, and various small indel mutations (6-203 bp deletions) in the target sequence were found. The fibroblasts mutated with the CRISPR/Cas9 system were applied for somatic cell nuclear transfer, and the reconstructed embryos were successfully developed into the blastocyst stage. In conclusion, the CRISPR/Cas9 system was successfully utilized in bovine cells and cloned embryos. This will be a useful technique to develop livestock transgenesis for agricultural science.
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
6-(Dimethylamino)purine, ≥98%